Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slfn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053687.1 | NP_446139.1 | 4 | 1725 | ||
XM_008767933.1 | XP_008766155.1 | 6 | 2187 | |||
GenBank mRNA | AF168795.1 | - | 4 | 1725 |
Gene Symbol: LOC103690169 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773862.1 | XP_008772084.1 | 2 | 855 |
Gene Symbol: Slfn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008768151.1 | XP_008766373.1 | 4 | 1491 |