Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MNT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020310.2 | NP_064706.1 | 2 | 722 | ||
XM_011523868.2 | XP_011522170.1 | 2 | 659 | |||
GenBank mRNA | AK291596.1 | - | 2 | 819 | ||
BC117563.1 | - | 2 | 472 | 3904263 | ||
X96401.1 | - | 2 | 682 |