Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other YWHAQ SIRNA Assays ›
          Gene Symbol:
          Ywhaq
          Gene Name:
          tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, theta polypeptide
          Gene Aliases:
          2700028P07Rik, AA409740, AU021156, R74690
          Chromosome Location:
          Chr.12: 21390329 - 21417436 on Build GRCm38
          UniGene ID:
          Mm.289630
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 70098

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Ywhaq
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_011739.3 NP_035869.1 2 698
          GenBank mRNA AK008948.1 - 3 468
          AK012300.1 - 2 431
          AK012540.1 - 3 433
          AK145568.1 - 3 423
          AK146371.1 - 3 423
          AK150850.1 - 3 423
          AK151437.1 - 3 472
          AK151715.1 - 3 451
          AK151864.1 - 3 472
          AK159403.1 - 3 456
          AK159817.1 - 3 472
          AK163237.1 - 3 425
          AK167397.1 - 3 424
          AK177592.1 - 2 183
          AK182050.1 - 2 185
          AK192231.1 - 2 183
          AK193357.1 - 2 183
          AK196448.1 - 2 183
          AK198944.1 - 2 183
          AK201174.1 - 2 184
          BC003837.1 - 2 211
          BC080802.1 - 3 413
          BC085299.1 - 3 478
          BC090838.1 - 2 698 6403848
          BC106164.1 - 2 400 3582392
          BC110680.1 - 2 407
          BC132252.1 - 2 467 40130529
          BC132530.1 - 2 467 40130807
          D87662.1 - 2 390
          U56243.1 - 2 373
          U57312.1 - 2 353

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: NM_006826, X56468, BC056867, X57347, BT020014, and others.
          This siRNA also targets the following transcripts in Rat: NM_013053, FQ218679, BC062409, D17614.

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          protein targeting
          signal transduction
          small GTPase mediated signal transduction
          negative regulation of ion transmembrane transport
          negative regulation of transcription, DNA-templated
          protein binding
          protein C-terminus binding
          protein domain specific binding
          ion channel binding
          protein N-terminus binding
          14-3-3 protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-gskvx:80/100.66.75.242:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0