Search
Search

Tktl1| Gene Symbol: Tktl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031379.2 | NP_113556.2 | 9, 10 | 1353 | ||
| GenBank mRNA | AF285571.1 | - | 9, 10 | 1328 | ||
| AJ627042.1 | - | 2, 3 | 243 | |||
| AK029546.1 | - | 9, 10 | 1354 | |||
| AK139841.1 | - | 9, 10 | 1353 | |||
| BC113768.1 | - | 9, 10 | 1328 | 40058456 | ||