Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other IL15 SIRNA Assays ›
          Gene Symbol:
          IL15
          Gene Name:
          interleukin 15
          Gene Aliases:
          IL-15
          Chromosome Location:
          Chr.4: 141636596 - 141733987 on Build GRCh38
          UniGene ID:
          Hs.168132
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 8746

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: IL15
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000585.4 NP_000576.1 8 793
          NM_172175.2 NP_751915.1 10 1114
          NR_037840.2 8 1269
          GenBank mRNA AK122993.1 - 7 5487
          AK290619.1 - 12 1086
          AY720442.1 - 5 349
          BC018149.2 - 8 1263 3851514
          BC100961.1 - 7 618 40004735
          BC100962.1 - 7 618 40004736
          BC100963.1 - 6 499 40004738
          CR541980.1 - 5 338
          CR542007.1 - 5 338
          U14407.1 - 8 735
          X94222.1 - 7 560
          X94223.1 - 7 560

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          interleukin superfamily

          Gene Ontology Categories:

          Function(s) Process(es)

          natural killer cell differentiation
          NK T cell proliferation
          inflammatory response
          immune response
          signal transduction
          cell-cell signaling
          aging
          positive regulation of cell proliferation
          skeletal muscle atrophy
          hyaluronan metabolic process
          positive regulation of interleukin-17 production
          positive regulation of natural killer cell proliferation
          positive regulation of natural killer cell differentiation
          positive regulation of tissue remodeling
          positive regulation of T cell proliferation
          tyrosine phosphorylation of Stat5 protein
          positive regulation of tyrosine phosphorylation of Stat3 protein
          extrathymic T cell selection
          regulation of T cell differentiation
          cell maturation
          lymph node development
          negative regulation of smooth muscle cell proliferation
          regulation of defense response to virus by host
          positive regulation of inflammatory response
          positive regulation of immune response
          cellular response to vitamin D
          positive regulation of protein O-linked glycosylation
          cytokine activity
          cytokine receptor binding
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0