Search
Search

Cntn4| Gene Symbol: Cntn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109749.1 | NP_001103219.1 | 11 | 1426 | ||
| NM_001109751.1 | NP_001103221.1 | 10 | 1514 | |||
| NM_173004.3 | NP_766592.2 | 11 | 1426 | |||
| GenBank mRNA | AK036262.1 | - | 11 | 1396 | ||
| AK043507.1 | - | 8 | 1371 | |||
| AK044694.1 | - | 1 | 174 | |||
| AK163520.1 | - | 8 | 1219 | |||
| AK173340.1 | - | 4 | 545 | |||
| BC046975.1 | - | 8 | 1048 | |||
| BC115766.1 | - | 11 | 1316 | 40060288 | ||