Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PSEN1 SIRNA Assays ›
          Gene Symbol:
          PSEN1
          Gene Name:
          presenilin 1
          Gene Aliases:
          AD3, FAD, PS-1, PS1, S182
          Chromosome Location:
          Chr.14: 73136435 - 73223691 on Build GRCh38
          UniGene ID:
          Hs.3260
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS108651
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PSEN1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000021.3 NP_000012.1 7 918
          NM_007318.2 NP_015557.2 7 906
          XM_005267864.2 XP_005267921.1 7 912
          XM_005267866.1 XP_005267923.1 7 901
          XM_011536971.2 XP_011535273.1 7 810
          XM_011536972.2 XP_011535274.1 7 1174
          XM_011536973.1 XP_011535275.1 7 1162
          XM_011536974.1 XP_011535276.1 7 803
          GenBank mRNA AF416717.1 - 5 634
          AK122722.1 - 5 2187
          AK223173.1 - 5 648
          AK312531.1 - 7 902
          BC011729.2 - 7 887 4423098
          KU178285.1 - 5 622
          KU178286.1 - 5 634
          L42110.1 - 7 882
          L76517.1 - 7 1175
          U40379.1 - 5 622
          U40380.1 - 5 622

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          aspartic protease

          Gene Ontology Categories:

          Function(s) Process(es)

          autophagosome assembly
          negative regulation of transcription from RNA polymerase II promoter
          activation of MAPKK activity
          blood vessel development
          cell fate specification
          somitogenesis
          neuron migration
          positive regulation of receptor recycling
          heart looping
          hematopoietic progenitor cell differentiation
          T cell activation involved in immune response
          neural retina development
          protein glycosylation
          membrane protein ectodomain proteolysis
          mitochondrial transport
          cellular response to DNA damage stimulus
          response to oxidative stress
          negative regulation of epidermal growth factor-activated receptor activity
          Notch signaling pathway
          Notch receptor processing
          memory
          post-embryonic development
          protein transport
          L-glutamate transport
          choline transport
          synaptic vesicle targeting
          single organismal cell-cell adhesion
          protein processing
          cerebral cortex cell migration
          Cajal-Retzius cell differentiation
          dorsal/ventral neural tube patterning
          embryonic limb morphogenesis
          positive regulation of proteasomal ubiquitin-dependent protein catabolic process
          endoplasmic reticulum calcium ion homeostasis
          beta-amyloid formation
          intracellular signal transduction
          regulation of phosphorylation
          amyloid precursor protein catabolic process
          myeloid dendritic cell differentiation
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          positive regulation of catalytic activity
          regulation of protein binding
          positive regulation of MAP kinase activity
          negative regulation of neuron apoptotic process
          skin morphogenesis
          positive regulation of transcription, DNA-templated
          regulation of synaptic plasticity
          thymus development
          neuron development
          skeletal system morphogenesis
          brain morphogenesis
          epithelial cell proliferation
          negative regulation of axonogenesis
          positive regulation of coagulation
          T cell receptor signaling pathway
          neuron apoptotic process
          negative regulation of ubiquitin-protein transferase activity
          smooth endoplasmic reticulum calcium ion homeostasis
          regulation of synaptic transmission, glutamatergic
          canonical Wnt signaling pathway
          regulation of resting membrane potential
          positive regulation of dendritic spine development
          calcium ion transmembrane transport
          negative regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          negative regulation of apoptotic signaling pathway
          endopeptidase activity
          aspartic-type endopeptidase activity
          calcium channel activity
          protein binding
          beta-catenin binding
          PDZ domain binding
          cadherin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0