Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TAP2 SIRNA Assays ›
          Gene Symbol:
          TAP2
          Gene Name:
          transporter 2, ATP binding cassette subfamily B member
          Gene Aliases:
          ABC18, ABCB3, APT2, D6S217E, PSF-2, PSF2, RING11
          Chromosome Location:
          Chr.6: 32821833 - 32838823 on Build GRCh38
          UniGene ID:
          Hs.502
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS110478
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TAP2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000544.3 NP_000535.3 2 513
          NM_001290043.1 NP_001276972.1 2 566
          NM_018833.2 NP_061313.2 2 513
          GenBank mRNA AB073779.1 - 1 391
          AB208953.1 - 2 460
          AF078671.1 - 2 470
          AF105151.1 - 2 470
          AK222823.1 - 1 422
          AK223300.1 - 2 473
          AK225657.1 - 2 494
          AK225693.1 - 2 473
          AK292963.1 - 2 496
          BC002751.2 - 1 399
          BG423357.1 - 2 513
          BT009906.1 - 1 391
          DC345324.1 - 2 566
          FM246456.1 - 1 391
          M74447.1 - 2 487
          M84748.1 - 2 419
          U07844.1 - 1 201
          Z22935.1 - 2 419
          Z22936.1 - 2 467

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ATP-binding cassette (ABC) transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          adaptive immune response
          antigen processing and presentation of peptide antigen via MHC class I
          antigen processing and presentation of exogenous protein antigen via MHC class Ib, TAP-dependent
          antigen processing and presentation of endogenous peptide antigen via MHC class I via ER pathway, TAP-dependent
          antigen processing and presentation of endogenous peptide antigen via MHC class Ib via ER pathway, TAP-dependent
          positive regulation of antigen processing and presentation of peptide antigen via MHC class I
          intracellular transport of viral protein in host cell
          antigen processing and presentation of endogenous peptide antigen via MHC class I
          cytosol to ER transport
          peptide antigen transport
          transmembrane transport
          transporter activity
          protein binding
          ATP binding
          peptide transporter activity
          peptide antigen-transporting ATPase activity
          MHC class Ib protein binding
          TAP1 binding
          tapasin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0