Search
Search

UCHL3| Gene Symbol: UCHL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270952.1 | NP_001257881.1 | 9 | 803 | ||
| NM_006002.4 | NP_005993.1 | 9 | 725 | |||
| XM_011535212.1 | XP_011533514.1 | 10 | 917 | |||
| XM_011535213.2 | XP_011533515.1 | 9 | 898 | |||
| XM_011535214.2 | XP_011533516.1 | 10 | 985 | |||
| XM_017020727.1 | XP_016876216.1 | 8 | 700 | |||
| GenBank mRNA | AA234624.1 | - | 2 | 93 | ||
| AK313665.1 | - | 9 | 715 | |||
| BC018125.2 | - | 9 | 684 | 3887201 | ||
| BT019359.1 | - | 9 | 654 | |||
| CR456855.1 | - | 9 | 654 | |||
| M30496.1 | - | 9 | 674 | |||