Search
Search

UCHL3| Gene Symbol: UCHL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270952.1 | NP_001257881.1 | 9 | 828 | ||
| NM_006002.4 | NP_005993.1 | 9 | 750 | |||
| XM_011535212.1 | XP_011533514.1 | 10 | 942 | |||
| XM_011535213.2 | XP_011533515.1 | 9 | 923 | |||
| XM_011535214.2 | XP_011533516.1 | 10 | 1010 | |||
| XM_017020727.1 | XP_016876216.1 | 8 | 725 | |||
| GenBank mRNA | AA234624.1 | - | 2 | 118 | ||
| AK313665.1 | - | 9 | 740 | |||
| BC018125.2 | - | 9 | 709 | 3887201 | ||
| BT019359.1 | - | 9 | 679 | |||
| M30496.1 | - | 9 | 699 | |||