Search
Search

PPFIA3| Gene Symbol: PPFIA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003660.3 | NP_003651.1 | 3 | 609 | ||
| NR_103842.1 | 3 | 609 | ||||
| XM_017027407.1 | XP_016882896.1 | 3 | 609 | |||
| GenBank mRNA | AB014554.2 | - | 3 | 592 | ||
| AK289757.1 | - | 3 | 314 | |||
| AK293798.1 | - | 1 | 60 | |||
| AK308483.1 | - | 1 | 100 | |||
| BC101518.1 | - | 3 | 335 | 8069024 | ||
| BC101520.1 | - | 3 | 335 | 8069026 | ||
| BC143470.1 | - | 3 | 335 | |||