Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MYT1L SIRNA Assays ›
          Gene Symbol:
          MYT1L
          Gene Name:
          myelin transcription factor 1 like
          Gene Aliases:
          MRD39, NZF1, ZC2H2C2, ZC2HC4B
          Chromosome Location:
          Chr.2: 1789113 - 2331388 on Build GRCh38
          UniGene ID:
          Hs.434418
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS118080
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MYT1L
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001303052.1 NP_001289981.1 24, 25 4285
          NM_015025.3 NP_055840.2 24, 25 4279
          XM_011510318.1 XP_011508620.1 25, 26 4416
          XM_011510319.1 XP_011508621.1 25, 26 4526
          XM_011510320.2 XP_011508622.1 25, 26 4457
          XM_011510321.2 XP_011508623.1 25, 26 4407
          XM_011510322.2 XP_011508624.1 25, 26 4220
          XM_011510323.1 XP_011508625.1 24, 25 4139
          XM_011510324.1 XP_011508626.1 25, 26 4410
          XM_011510325.1 XP_011508627.1 25, 26 4410
          XM_011510326.1 XP_011508628.1 24, 25 4353
          XM_017003604.1 XP_016859093.1 26, 27 4505
          XM_017003605.1 XP_016859094.1 25, 26 4084
          XM_017003606.1 XP_016859095.1 25, 26 4520
          XM_017003607.1 XP_016859096.1 25, 26 4451
          XM_017003609.1 XP_016859098.1 24, 25 4332
          XM_017003610.1 XP_016859099.1 25, 26 4431
          XM_017003611.1 XP_016859100.1 24, 25 4347
          XM_017003612.1 XP_016859101.1 24, 25 4457
          XM_017003613.1 XP_016859102.1 24, 25 4451
          XM_017003614.1 XP_016859103.1 24, 25 4290
          XM_017003620.1 XP_016859109.1 18, 19 3406
          GenBank mRNA AB029029.2 - 25, 26 4064
          AF036943.1 - 24, 25 4180
          BC071612.1 - 6, 7 768
          BC137272.1 - 24, 25 4082 9021269
          BC137273.1 - 24, 25 4082 9021270
          BC150281.1 - 25, 26 4064 9007267

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          nervous system development
          cell differentiation
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          zinc ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-g9g6n:80/100.66.79.69:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0