Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAALADL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207015.2 | NP_996898.2 | 6 | 1207 | ||
XM_006713560.2 | XP_006713623.1 | 8 | 1290 | |||
XM_011512612.2 | XP_011510914.1 | 6 | 4544 | |||
XM_011512613.1 | XP_011510915.1 | 6 | 1352 | |||
XM_011512616.2 | XP_011510918.1 | 5 | 2082 | |||
XM_011512617.2 | XP_011510919.1 | 6 | 4542 | |||
XM_017006070.1 | XP_016861559.1 | 10 | 1732 | |||
XM_017006071.1 | XP_016861560.1 | 11 | 1802 | |||
XM_017006072.1 | XP_016861561.1 | 12 | 1936 | |||
XM_017006073.1 | XP_016861562.1 | 10 | 1535 | |||
XM_017006074.1 | XP_016861563.1 | 12 | 3954 | |||
XM_017006075.1 | XP_016861564.1 | 9 | 2679 | |||
XM_017006076.1 | XP_016861565.1 | 9 | 1472 | |||
XM_017006077.1 | XP_016861566.1 | 11 | 2883 | |||
XM_017006078.1 | XP_016861567.1 | 9 | 2700 | |||
XM_017006079.1 | XP_016861568.1 | 7 | 2176 | |||
XM_017006080.1 | XP_016861569.1 | 7 | 2547 | |||
XM_017006081.1 | XP_016861570.1 | 6 | 4544 | |||
XM_017006083.1 | XP_016861572.1 | 6 | 836 | |||
GenBank mRNA | AJ607397.1 | - | 2 | 150 | ||
AK308416.1 | - | 6 | 1223 | |||
AL832144.1 | - | 6 | 1207 | |||
AL832931.1 | - | 3 | 292 |