Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FGF1 SIRNA Assays ›
Gene Symbol:
FGF1
Gene Name:
fibroblast growth factor 1
Gene Aliases:
AFGF, ECGF, ECGF-beta, ECGFA, ECGFB, FGF-1, FGF-alpha, FGFA, GLIO703, HBGF-1, HBGF1
Chromosome Location:
Chr.5: 142592178 - 142698070 on Build GRCh38
UniGene ID:
Hs.483635
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS142001
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FGF1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000800.4
NP_000791.1
4
895
NM_001144892.2
NP_001138364.1
4
415
NM_001144934.1
NP_001138406.1
5
608
NM_001144935.1
NP_001138407.1
4
514
NM_001257205.1
NP_001244134.1
4
805
NM_001257206.1
NP_001244135.1
4
802
NM_001257207.1
NP_001244136.1
5
548
NM_001257208.1
NP_001244137.1
4
546
NM_001257209.1
NP_001244138.1
4
561
NM_001257210.1
NP_001244139.1
4
545
NM_001257211.1
NP_001244140.1
4
543
NM_001257212.1
NP_001244141.1
4
412
NM_033136.3
NP_149127.1
3
791
NM_033137.2
NP_149128.1
2
249
XM_005268390.4
XP_005268447.1
4
485
XM_011537607.2
XP_011535909.1
5
605
GenBank mRNA
AK289762.1
-
4
742
AK312301.1
-
5
548
BC032697.1
-
4
412
5403677
BG706412.1
-
4
470
DA111414.1
-
4
433
DA189432.1
-
4
525
DA228737.1
-
3
362
DA326831.1
-
3
449
DA388578.1
-
4
508
DA389088.1
-
4
458
DA575048.1
-
4
412
DA660034.1
-
4
545
DQ655917.2
-
4
787
M13361.1
-
3
360
S67291.1
-
3
322
S67292.1
-
2
218
S67294.1
-
2
215
X51943.1
-
3
356
X65778.1
-
3
332
X65779.1
-
2
228
Back To Top
More Information
Panther Classification:
Molecular Function -
growth factor
intercellular signal molecule
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
activation of MAPK activity
angiogenesis
organ induction
signal transduction
multicellular organism development
cell proliferation
positive regulation of cell proliferation
fibroblast growth factor receptor signaling pathway
anatomical structure morphogenesis
positive regulation of endothelial cell migration
regulation of phosphatidylinositol 3-kinase signaling
peptidyl-tyrosine phosphorylation
cell differentiation
lung development
positive regulation of cell migration
activation of protein kinase B activity
cellular response to heat
phosphatidylinositol-3-phosphate biosynthetic process
positive regulation of MAP kinase activity
positive regulation of GTPase activity
positive regulation of cholesterol biosynthetic process
positive regulation of angiogenesis
positive regulation of transcription from RNA polymerase II promoter
phosphatidylinositol phosphorylation
phosphatidylinositol-mediated signaling
positive regulation of epithelial cell proliferation
positive regulation of cell division
branch elongation involved in ureteric bud branching
positive regulation of ERK1 and ERK2 cascade
mesonephric epithelium development
regulation of endothelial tube morphogenesis
positive regulation of intracellular signal transduction
positive regulation of sprouting angiogenesis
regulation of endothelial cell chemotaxis to fibroblast growth factor
protein tyrosine kinase activity
Ras guanyl-nucleotide exchange factor activity
fibroblast growth factor receptor binding
integrin binding
protein binding
growth factor activity
heparin binding
1-phosphatidylinositol-3-kinase activity
Hsp70 protein binding
S100 protein binding
phosphatidylinositol-4,5-bisphosphate 3-kinase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0