Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAUS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015302.1 | NP_056117.1 | 17 | 1647 | ||
XM_011526684.2 | XP_011524986.1 | 16 | 1534 | |||
GenBank mRNA | AB020648.1 | - | 17 | 1620 | ||
BC013947.2 | - | 17 | 1619 | |||
BC064390.1 | - | 15 | 1836 | |||
BC157882.1 | - | 17 | 1638 | 8862704 | ||
BC171905.1 | - | 16 | 1892 |