Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TVP23B SIRNA Assays ›
          Gene Symbol:
          TVP23B MKL2 TVP23C TVP23C-CDRT4
          Gene Name:
          trans-golgi network vesicle protein 23 homolog B (S. cerevisiae)
          MKL1/myocardin like 2
          trans-golgi network vesicle protein 23 homolog C (S. cerevisiae)
          TVP23C-CDRT4 readthrough
          Gene Aliases:
          CGI-148, FAM18B, FAM18B1, NPD008, YDR084C
          MRTF-B, MRTFB, NPD001
          FAM18B2
          FAM18B2-CDRT4
          Chromosome Location:
          Chr.17: 18780995 - 18806713 on Build GRCh38
          Chromosome Location:
          Chr.16: 14071076 - 14266779 on Build GRCh38
          Chromosome Location:
          Chr.17: 15502264 - 15563631 on Build GRCh38
          Chromosome Location:
          Chr.17: 15436015 - 15563631 on Build GRCh38
          UniGene ID:
          Hs.87295
          Hs.49143
          Hs.164595
          Hs.164595
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          siRNA ID HSS147245
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price 459.00
          Your Price
          Online offer:
          459.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TVP23B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001316919.1 NP_001303848.1 2 1267
          NM_001316920.1 NP_001303849.1 2 48
          NM_001316921.1 NP_001303850.1 2 337
          NM_001316922.1 NP_001303851.1 2 342
          NM_001316923.1 NP_001303852.1 2 342
          NM_001316924.1 NP_001303853.1 2 342
          NM_016078.5 NP_057162.4 2 342
          GenBank mRNA AF151906.1 - 2 342
          AF223467.1 - 2 342
          AK290813.1 - 2 66
          BC008430.1 - 2 68 4292664
          BQ433266.1 - 1 43
          BX537929.1 - 2 58
          BX648167.1 - 2 1267
          CN390782.1 - 1 40
          DA067248.1 - 2 48
          DA551966.1 - 2 63
          DA773045.1 - 2 67
          DB143840.1 - 2 310
          Gene Symbol: MKL2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA BC047761.1 - 1 70
          Gene Symbol: TVP23C
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001135036.1 NP_001128508.1 2 226
          NM_145301.2 NP_660344.2 2 226
          GenBank mRNA AK310363.1 - 2 185
          DC393880.1 - 2 227
          Gene Symbol: TVP23C-CDRT4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001204478.1 NP_001191407.1 2 226
          NR_037924.1 2 226
          GenBank mRNA AB074181.1 - 2 156
          AK303475.1 - 2 154
          DA796743.1 - 2 67

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor

          Gene Ontology Categories:

          Function(s)

          protein secretion
          vesicle-mediated transport

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0