Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SLFNL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168247.2 | NP_001161719.1 | 3 | 814 | ||
NM_001300859.1 | NP_001287788.1 | 3 | 721 | |||
NM_144990.3 | NP_659427.3 | 3 | 658 | |||
XM_005270597.2 | XP_005270654.1 | 2 | 1397 | |||
XM_005270598.2 | XP_005270655.1 | 2 | 713 | |||
XM_005270599.2 | XP_005270656.1 | 3 | 721 | |||
XM_006710432.3 | XP_006710495.1 | 2 | 1808 | |||
XM_011540943.1 | XP_011539245.1 | 2 | 1344 | |||
XM_011540945.1 | XP_011539247.1 | 2 | 1281 | |||
XM_011540947.1 | XP_011539249.1 | 2 | 806 | |||
XM_011540948.1 | XP_011539250.1 | 3 | 1259 | |||
XM_011540949.1 | XP_011539251.1 | 2 | 1189 | |||
XM_011540950.1 | XP_011539252.1 | 2 | 1159 | |||
XM_011540951.1 | XP_011539253.1 | 3 | 713 | |||
XM_011540952.2 | XP_011539254.1 | 2 | 1136 | |||
XM_011540953.2 | XP_011539255.1 | 2 | 1716 | |||
XM_011540954.1 | XP_011539256.1 | 2 | 1067 | |||
XM_017000573.1 | XP_016856062.1 | 2 | 1397 | |||
XM_017000574.1 | XP_016856063.1 | 2 | 1397 | |||
GenBank mRNA | AK097609.1 | - | 3 | 709 | ||
AK292296.1 | - | 3 | 784 | |||
BC022037.2 | - | 3 | 620 | 4826722 | ||
BC037879.1 | - | 1 | 1470 | 5274032 | ||
BC050353.1 | - | 1 | 2978 | 5265392 |