Search
Search

FDPS| Gene Symbol: FDPS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135821.1 | NP_001129293.1 | 3 | 378 | ||
| NM_001135822.1 | NP_001129294.1 | 2 | 235 | |||
| NM_001242824.1 | NP_001229753.1 | 2 | 256 | |||
| NM_002004.3 | NP_001995.1 | 3 | 433 | |||
| XM_005244962.1 | XP_005245019.1 | 2 | 146 | |||
| XM_005244963.1 | XP_005245020.1 | 2 | 201 | |||
| GenBank mRNA | AK021828.1 | - | 1 | 1169 | ||
| AK022841.1 | - | 1 | 1144 | |||
| AK291084.1 | - | 3 | 234 | |||
| BC010004.2 | - | 2 | 222 | 4132071 | ||
| BP370037.1 | - | 2 | 260 | |||
| CN346026.1 | - | 2 | 205 | |||
| D14697.1 | - | 3 | 328 | |||
| DA610405.1 | - | 2 | 201 | |||
| J05262.1 | - | 1 | 22 | |||