Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PPP1R15A SIRNA Assays ›
          Gene Symbol:
          PPP1R15A
          Gene Name:
          protein phosphatase 1 regulatory subunit 15A
          Gene Aliases:
          GADD34
          Chromosome Location:
          Chr.19: 48872392 - 48876062 on Build GRCh38
          UniGene ID:
          Hs.631593
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS177543
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PPP1R15A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_014330.3 NP_055145.3 2 790
          GenBank mRNA AK001361.1 - 2 761
          AK225632.1 - 2 761
          AK297615.1 - 3 678
          AK298765.1 - 2 75
          AK311366.1 - 2 315
          BC003067.2 - 2 743 3507556
          CR457259.1 - 1 521
          U83981.1 - 2 743

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of protein phosphorylation
          apoptotic process
          cellular response to DNA damage stimulus
          cell cycle arrest
          aging
          negative regulation of cell proliferation
          negative regulation of peptidyl-threonine phosphorylation
          negative regulation of protein processing
          endoplasmic reticulum unfolded protein response
          positive regulation of translational initiation in response to stress
          negative regulation of phosphoprotein phosphatase activity
          positive regulation of phosphoprotein phosphatase activity
          cellular response to UV
          response to endoplasmic reticulum stress
          negative regulation of protein dephosphorylation
          cellular response to drug
          response to immobilization stress
          regulation of translational initiation by eIF2 alpha dephosphorylation
          response to cocaine
          positive regulation of apoptotic process
          regulation of angiogenesis
          negative regulation of cell death
          regulation of endoplasmic reticulum stress-induced eIF2 alpha phosphorylation
          intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
          peptidyl-serine dephosphorylation
          protein localization to endoplasmic reticulum
          cellular response to antibiotic
          cellular response to carbohydrate stimulus
          cellular response to methyl methanesulfonate
          cellular response to calcium ion starvation
          response to environmental enrichment
          cellular response to oxygen-glucose deprivation
          positive regulation of peptidyl-serine dephosphorylation
          negative regulation of PERK-mediated unfolded protein response
          negative regulation of endoplasmic reticulum stress-induced eIF2 alpha phosphorylation
          positive regulation of endoplasmic reticulum stress-induced eIF2 alpha dephosphorylation
          cellular response to desipramine
          cellular response to cordycepin
          cellular response to gold(3+)
          cellular response to methamphetamine hydrochloride
          negative regulation of transcription from RNA polymerase II promoter in response to endoplasmic reticulum stress
          protein binding
          protein phosphatase 1 binding
          protein phosphatase type 1 regulator activity
          protein kinase binding
          protein phosphatase type 1 activator activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-gk9w8:80/100.66.76.60:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0