Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BLOC1S2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001342.2 | NP_001001342.1 | 2 | 325 | ||
NM_001282436.1 | NP_001269365.1 | 4 | 390 | |||
NM_001282437.1 | NP_001269366.1 | 3 | 401 | |||
NM_001282438.1 | NP_001269367.1 | 3 | 252 | |||
NM_001282439.1 | NP_001269368.1 | 3 | 282 | |||
NM_173809.4 | NP_776170.2 | 3 | 282 | |||
XM_017016102.1 | XP_016871591.1 | 3 | 381 | |||
GenBank mRNA | AB593084.1 | - | 3 | 244 | ||
AK054697.1 | - | 2 | 318 | |||
AK298969.1 | - | 3 | 250 | |||
AY278457.1 | - | 2 | 280 | |||
AY531265.1 | - | 3 | 229 | |||
BC020494.1 | - | 2 | 153 | 3900723 | ||
BF666010.1 | - | 4 | 357 | |||
BG545838.1 | - | 2 | 320 | |||
BG679165.1 | - | 3 | 254 | |||
BI753469.1 | - | 3 | 242 | |||
CN340962.1 | - | 3 | 369 | |||
DB456773.1 | - | 3 | 250 | |||
HY107885.1 | - | 2 | 327 | |||
HY165508.1 | - | 3 | 253 |