Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GYG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184720.1 | NP_001171649.1 | 4 | 619 | ||
NM_001184721.1 | NP_001171650.1 | 4 | 619 | |||
NM_004130.3 | NP_004121.2 | 4 | 619 | |||
XM_017006275.1 | XP_016861764.1 | 3 | 376 | |||
GenBank mRNA | AF087942.1 | - | 4 | 439 | ||
AK299962.1 | - | 4 | 515 | |||
AK312283.1 | - | 4 | 455 | |||
BC000033.2 | - | 4 | 475 | 3504538 | ||
BC031096.2 | - | 3 | 380 | |||
BG702275.1 | - | 4 | 622 | |||
CR536547.1 | - | 3 | 386 | |||
U31525.1 | - | 4 | 513 | |||
U44131.1 | - | 3 | 394 | |||
X79537.1 | - | 4 | 469 |