Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FAM49B SIRNA Assays ›
Gene Symbol:
FAM49B
Gene Name:
family with sequence similarity 49 member B
Gene Aliases:
BM-009, L1
Chromosome Location:
Chr.8: 129839593 - 130016697 on Build GRCh38
UniGene ID:
Hs.126941
Species:
Human
siRNA ID
HSS182127
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FAM49B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001256763.1
NP_001243692.1
6
620
NM_016623.4
NP_057707.3
5
533
NR_046359.1
8
886
NR_046360.1
8
796
NR_046361.1
7
786
XM_005250966.3
XP_005251023.1
4
389
XM_011517108.2
XP_011515410.1
7
682
XM_011517109.2
XP_011515411.1
7
454
XM_011517110.2
XP_011515412.1
6
594
XM_011517111.2
XP_011515413.1
6
412
XM_011517112.2
XP_011515414.1
8
637
XM_011517113.2
XP_011515415.1
7
648
XM_011517114.2
XP_011515416.1
7
554
XM_011517115.2
XP_011515417.1
6
561
XM_011517116.2
XP_011515418.1
7
595
XM_011517117.2
XP_011515419.1
6
564
XM_011517118.2
XP_011515420.1
6
508
XM_011517120.2
XP_011515422.1
5
507
XM_011517121.2
XP_011515423.1
8
519
XM_011517122.2
XP_011515424.1
6
598
XM_011517123.2
XP_011515425.1
6
371
XM_011517124.2
XP_011515426.1
5
486
XM_011517125.2
XP_011515427.1
5
425
XM_017013539.1
XP_016869028.1
8
641
XM_017013540.1
XP_016869029.1
8
500
XM_017013541.1
XP_016869030.1
6
628
XM_017013542.1
XP_016869031.1
8
678
XM_017013543.1
XP_016869032.1
8
778
XM_017013544.1
XP_016869033.1
8
560
XM_017013545.1
XP_016869034.1
9
824
XM_017013546.1
XP_016869035.1
7
591
XM_017013547.1
XP_016869036.1
9
683
XM_017013548.1
XP_016869037.1
7
687
XM_017013549.1
XP_016869038.1
7
550
XM_017013550.1
XP_016869039.1
6
601
XM_017013551.1
XP_016869040.1
8
596
XM_017013552.1
XP_016869041.1
8
641
XM_017013553.1
XP_016869042.1
7
454
XM_017013554.1
XP_016869043.1
9
666
XM_017013555.1
XP_016869044.1
6
621
XM_017013556.1
XP_016869045.1
6
367
XM_017013557.1
XP_016869046.1
7
413
XM_017013558.1
XP_016869047.1
7
558
XM_017013559.1
XP_016869048.1
7
417
XM_017013560.1
XP_016869049.1
5
532
XM_017013561.1
XP_016869050.1
5
284
XM_017013562.1
XP_016869051.1
6
471
GenBank mRNA
AF208851.1
-
7
472
AK001226.1
-
1
39
AK002059.1
-
8
596
AK130372.1
-
5
420
AY364259.1
-
2
87
AY598320.1
-
3
219
BC003599.1
-
6
399
3501828
BC016345.1
-
7
540
4041187
BC017297.2
-
8
768
5017493
BX419336.2
-
6
621
CR749628.1
-
6
366
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0