Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsga10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290720.1 | NP_001277649.1 | 14 | 1897 | ||
NM_001290721.1 | NP_001277650.1 | 13 | 1704 | |||
NM_207228.3 | NP_997111.1 | 14 | 1764 | |||
GenBank mRNA | AF530050.3 | - | 13 | 1709 | ||
AK030254.1 | - | 14 | 1897 | |||
AY618264.1 | - | 13 | 1709 | |||
BC053407.1 | - | 12 | 1532 | |||
BC066782.1 | - | 14 | 1761 | 6705127 | ||
CK789086.1 | - | 3 | 273 |