Search
Search

Arhgap39| Gene Symbol: Arhgap39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168288.1 | NP_001161760.1 | 3 | 394 | ||
| NM_198420.2 | NP_940812.2 | 3 | 394 | |||
| GenBank mRNA | AK035479.1 | - | 2 | 99 | ||
| AK141623.1 | - | 3 | 354 | |||
| AK173232.1 | - | 3 | 329 | |||
| BC060637.1 | - | 3 | 375 | 6849400 | ||
| CJ131891.1 | - | 3 | 394 | |||