Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001160.3 | NP_001001160.1 | 11 | 2793 | ||
NM_001289674.1 | NP_001276603.1 | 10 | 2624 | |||
NM_001289675.1 | NP_001276604.1 | 10 | 2618 | |||
GenBank mRNA | AK129466.1 | - | 9 | 2170 | ||
AK147230.1 | - | 11 | 2685 | |||
AK147681.1 | - | 10 | 2483 | |||
BC061475.1 | - | 8 | 1265 | |||
BC070445.1 | - | 10 | 2624 | 30536738 |