Search
Search

Gipc2| Gene Symbol: Gipc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016867.1 | NP_058563.1 | 5, 6 | 798 | ||
| GenBank mRNA | AF061262.1 | - | 5, 6 | 798 | ||
| AK008307.1 | - | 5, 6 | 615 | |||
| AK008626.1 | - | 5, 6 | 811 | |||
| AK168560.1 | - | 5, 6 | 809 | |||
| BC013440.1 | - | 5, 6 | 852 | 4238716 | ||