Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fads1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146094.2 | NP_666206.1 | 5 | 809 | ||
GenBank mRNA | AB072976.1 | - | 5 | 724 | ||
AF061504.1 | - | 5 | 711 | |||
AK033308.1 | - | 3 | 166 | |||
AK080706.1 | - | 5 | 713 | |||
AK083959.1 | - | 5 | 671 | |||
AK154367.1 | - | 5 | 759 | |||
AK210021.1 | - | 3 | 301 | |||
BC022139.1 | - | 5 | 754 | 5100199 | ||
BC026831.1 | - | 5 | 776 | 4239695 | ||
BC026848.1 | - | 5 | 749 | 4196030 | ||
BC063053.1 | - | 5 | 669 | 5696722 | ||
KF987079.1 | - | 5 | 693 |