Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cept1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293694.1 | NP_001280623.1 | 8 | 1240 | ||
NM_133869.4 | NP_598630.2 | 8 | 1240 | |||
GenBank mRNA | AK037116.1 | - | 8 | 1200 | ||
BC021753.1 | - | 8 | 1169 | 4167222 | ||
BC023783.1 | - | 8 | 1180 | 5346896 |