Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ogdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252282.1 | NP_001239211.1 | 18 | 2379 | ||
NM_001252283.1 | NP_001239212.1 | 18 | 2367 | |||
NM_001252287.1 | NP_001239216.1 | 17 | 2410 | |||
NM_001252288.1 | NP_001239217.1 | 17 | 2382 | |||
NM_010956.4 | NP_035086.2 | 17 | 2334 | |||
GenBank mRNA | AK147289.1 | - | 17 | 2335 | ||
AK150009.1 | - | 17 | 2300 | |||
AK169286.1 | - | 17 | 2326 | |||
AK220536.1 | - | 18 | 2341 | |||
BC013670.1 | - | 17 | 2268 | 4188988 | ||
BC025040.1 | - | 2 | 134 | |||
BC029143.1 | - | 2 | 144 | |||
BC031165.1 | - | 2 | 113 | |||
BC049104.1 | - | 17 | 2335 | 6417392 | ||
BC057354.1 | - | 18 | 2295 | 6827509 |