Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ip6k1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013785.2 | NP_038813.2 | 3 | 557 | ||
GenBank mRNA | AF177144.1 | - | 2 | 257 | ||
AK088872.1 | - | 3 | 574 | |||
AK166143.1 | - | 3 | 561 | |||
AK171310.1 | - | 3 | 562 | |||
AK172921.1 | - | 3 | 616 | |||
AK180416.1 | - | 2 | 74 | |||
AK184546.1 | - | 2 | 74 | |||
AK202332.1 | - | 2 | 98 | |||
BC059006.1 | - | 3 | 557 | 6827203 |