Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp672 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256516.1 | NP_001243445.1 | 4 | 597 | ||
NM_001256517.1 | NP_001243446.1 | 4 | 566 | |||
NM_001256518.1 | NP_001243447.1 | 5 | 848 | |||
NM_001256519.1 | NP_001243448.1 | 6 | 777 | |||
NM_001256520.1 | NP_001243449.1 | 3 | 468 | |||
NM_178761.5 | NP_848876.2 | 3 | 436 | |||
GenBank mRNA | AK029711.1 | - | 5 | 853 | ||
AK035113.1 | - | 3 | 415 | |||
AK085166.1 | - | 4 | 526 | |||
BC003258.1 | - | 1 | 239 | 3500812 | ||
BC034706.1 | - | 4 | 544 | 3582315 | ||
BU148968.1 | - | 4 | 529 | |||
CA321162.1 | - | 4 | 530 | |||
CV562361.1 | - | 3 | 460 | |||
CV783882.1 | - | 6 | 726 |