Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpd52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025261.1 | NP_001020432.1 | 8 | 781 | ||
NM_001025262.1 | NP_001020433.1 | 6 | 712 | |||
NM_001025263.1 | NP_001020434.1 | 7 | 657 | |||
NM_001025264.1 | NP_001020435.1 | 7 | 642 | |||
NM_009412.2 | NP_033438.1 | 6 | 615 | |||
GenBank mRNA | AK005456.1 | - | 6 | 596 | ||
AK032111.1 | - | 7 | 620 | |||
AK150935.1 | - | 6 | 578 | |||
AK151471.1 | - | 6 | 576 | |||
AK151744.1 | - | 6 | 576 | |||
AK152543.1 | - | 6 | 576 | |||
AK153266.1 | - | 6 | 576 | |||
AK153292.1 | - | 6 | 586 | |||
AK161753.1 | - | 6 | 577 | |||
AK168379.1 | - | 6 | 571 | |||
AK168817.1 | - | 6 | 579 | |||
AK169751.1 | - | 6 | 608 | |||
AK170725.1 | - | 6 | 561 | |||
AK171050.1 | - | 6 | 556 | |||
AK179073.1 | - | 1 | 59 | |||
AK181915.1 | - | 2 | 143 | |||
AK190065.1 | - | 1 | 59 | |||
AK198625.1 | - | 2 | 143 | |||
AK218627.1 | - | 2 | 144 | |||
AY048852.1 | - | 6 | 704 | |||
BC002036.1 | - | 6 | 595 | 3593963 | ||
BC004068.1 | - | 6 | 548 | 3590969 | ||
BC094018.1 | - | 6 | 571 | 3674995 | ||
U44426.1 | - | 6 | 544 |