Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scara5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168318.1 | NP_001161790.1 | 4 | 872 | ||
NM_028903.2 | NP_083179.2 | 4 | 872 | |||
GenBank mRNA | AK016543.1 | - | 4 | 907 | ||
AK016912.1 | - | 4 | 839 | |||
AK033154.1 | - | 4 | 839 | |||
AK133176.1 | - | 4 | 873 | |||
BC016096.1 | - | 4 | 848 | 4506472 | ||
BC023907.1 | - | 4 | 788 | 5321978 | ||
BC032159.1 | - | 4 | 828 | 5354657 | ||
DQ122126.1 | - | 3 | 522 |