Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddit4l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030143.4 | NP_084419.2 | 2 | 337 | ||
GenBank mRNA | AF335325.1 | - | 1 | 137 | ||
AK006617.1 | - | 2 | 336 | |||
AK018944.1 | - | 2 | 339 | |||
AK053333.1 | - | 2 | 275 | |||
AK075693.1 | - | 2 | 226 | |||
BC038131.1 | - | 2 | 236 | |||
BC147181.1 | - | 1 | 116 | 9056209 | ||
BC147182.1 | - | 1 | 116 | 9056220 |