Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HYAL1 SIRNA Assays ›
Gene Symbol:
Hyal1
Gene Name:
hyaluronoglucosaminidase 1
Gene Aliases:
-
Chromosome Location:
Chr.8: 116332924 - 116335237 on Build Rnor_6.0
UniGene ID:
Rn.124438
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
RSS327962
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1330001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Hyal1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_207616.1
NP_997499.1
2
814
GenBank mRNA
AB100600.1
-
2
814
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Human: NM_153285, NM_153282, NM_033159, NM_153281, NM_153283, and others.
Panther Classification:
Molecular Function -
glycosidase
Gene Ontology Categories:
Function(s)
Process(es)
response to reactive oxygen species
carbohydrate metabolic process
inflammatory response
response to virus
positive regulation of epithelial cell migration
hyaluronan metabolic process
hyaluronan biosynthetic process
hyaluronan catabolic process
positive regulation of cell growth
negative regulation of cell growth
cellular response to platelet-derived growth factor stimulus
cellular response to fibroblast growth factor stimulus
positive regulation of angiogenesis
positive regulation of cell adhesion
positive regulation of growth
response to antibiotic
positive regulation of epithelial cell proliferation
cartilage development
embryonic skeletal joint morphogenesis
cellular response to interleukin-1
cellular response to tumor necrosis factor
cellular response to pH
cellular response to UV-B
positive regulation of hyaluranon cable assembly
virus receptor activity
hyalurononglucosaminidase activity
transcription factor binding
hyaluronan synthase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-kfbvh:80/100.66.79.53:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0