Search
Search

| Gene Symbol: RT1-Db1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | X53054.1 | - | 2 | 271 | ||
| Gene Symbol: RT1-Bb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004084.2 | NP_001004084.1 | 2 | 324 | ||
| GenBank mRNA | AY626187.1 | - | 2 | 256 | ||
| BC079121.1 | - | 2 | 309 | 7126686 | ||
| KC222940.1 | - | 2 | 256 | |||