Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mief2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006246545.2 | XP_006246607.1 | 4 | 1603 | ||
XM_008767888.1 | XP_008766110.1 | 3 | 3012 | |||
XM_008767889.1 | XP_008766111.1 | 4 | 3661 | |||
XM_573103.6 | XP_573103.3 | 4 | 2192 |