Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BIN1 SIRNA Assays ›
Gene Symbol:
BIN1
Gene Name:
bridging integrator 1
Gene Aliases:
AMPH2, AMPHL, SH3P9
Chromosome Location:
Chr.2: 127048023 - 127107400 on Build GRCh38
UniGene ID:
Hs.193163
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s1342
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BIN1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001320632.1
NP_001307561.1
4, 5
697
NM_001320633.1
NP_001307562.1
4, 5
697
NM_001320634.1
NP_001307563.1
4, 5
463
NM_001320640.1
NP_001307569.1
4, 5
697
NM_001320641.1
NP_001307570.1
4, 5
697
NM_001320642.1
NP_001307571.1
4, 5
1191
NM_004305.3
NP_004296.1
4, 5
697
NM_139343.2
NP_647593.1
4, 5
697
NM_139344.2
NP_647594.1
4, 5
697
NM_139345.2
NP_647595.1
4, 5
697
NM_139346.2
NP_647596.1
4, 5
697
NM_139347.2
NP_647597.1
4, 5
697
NM_139348.2
NP_647598.1
4, 5
697
NM_139349.2
NP_647599.1
4, 5
697
NM_139350.2
NP_647600.1
4, 5
697
NM_139351.2
NP_647601.1
4, 5
697
XM_005263642.3
XP_005263699.1
4, 5
770
XM_005263643.3
XP_005263700.1
4, 5
770
XM_005263645.2
XP_005263702.1
4, 5
254
XM_006712425.3
XP_006712488.1
4, 5
770
XM_006712426.3
XP_006712489.1
4, 5
770
XM_006712427.2
XP_006712490.1
4, 5
254
XM_006712428.3
XP_006712491.1
4, 5
770
XM_006712429.3
XP_006712492.1
4, 5
770
XM_006712431.3
XP_006712494.1
4, 5
770
XM_006712432.3
XP_006712495.1
4, 5
770
XM_006712433.3
XP_006712496.1
4, 5
770
XM_006712434.3
XP_006712497.1
4, 5
770
XM_011510975.2
XP_011509277.1
4, 5
1403
XM_017003819.1
XP_016859308.1
4, 5
584
XM_017003820.1
XP_016859309.1
4, 5
770
XM_017003821.1
XP_016859310.1
4, 5
770
XM_017003822.1
XP_016859311.1
4, 5
770
XM_017003823.1
XP_016859312.1
4, 5
254
XM_017003824.1
XP_016859313.1
4, 5
254
XM_017003825.1
XP_016859314.1
4, 5
584
XM_017003826.1
XP_016859315.1
4, 5
254
XM_017003827.1
XP_016859316.1
4, 5
770
XM_017003828.1
XP_016859317.1
4, 5
770
GenBank mRNA
AF001383.1
-
4, 5
484
AF004015.1
-
4, 5
313
AF043898.1
-
4, 5
313
AF043899.1
-
4, 5
313
AF043900.1
-
4, 5
313
AF043901.1
-
4, 5
313
AF068914.1
-
4, 5
373
AF068915.1
-
4, 5
373
AF068916.1
-
4, 5
373
AF068917.1
-
4, 5
373
AF068918.1
-
4, 5
373
AK295300.1
-
4, 5
415
AK301153.1
-
4, 5
463
AL537027.3
-
4, 5
451
AL713697.1
-
4, 5
658
BC004101.2
-
4, 5
373
3688734
BM742027.1
-
4, 5
415
BT006865.1
-
4, 5
313
U68485.1
-
4, 5
359
U87558.1
-
4, 5
334
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
endocytosis
nucleus organization
cell proliferation
viral process
regulation of endocytosis
muscle cell differentiation
positive regulation of apoptotic process
regulation of neuron differentiation
positive regulation of astrocyte differentiation
lipid tube assembly
regulation of cell cycle arrest
protein binding
identical protein binding
tau protein binding
actin filament binding
RNA polymerase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System