Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC103692142 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762959.1 | XP_008761181.1 | 3 | 416 |
Gene Symbol: Hoxa13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271355.1 | NP_001258284.1 | 2 | 1021 |
Gene Symbol: LOC100910298 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749763.2 | XP_003749811.1 | 2 | 632 |