Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp3a73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006248850.2 | XP_006248912.1 | 5 | 1599 | ||
XM_008769027.1 | XP_008767249.1 | 5 | 1599 | |||
XM_008769028.1 | XP_008767250.1 | 4 | 1028 | |||
XM_008769029.1 | XP_008767251.1 | 5 | 705 |