Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278317.1 | NP_001265246.1 | 3 | 912 | ||
NM_012304.4 | NP_036436.1 | 3 | 1144 | |||
XM_011513998.1 | XP_011512300.1 | 3 | 703 | |||
XM_017009262.1 | XP_016864751.1 | 3 | 5124 | |||
GenBank mRNA | AB020647.2 | - | 3 | 1144 | ||
AF174593.1 | - | 2 | 639 | |||
AF199356.1 | - | 3 | 667 | |||
AK309585.1 | - | 3 | 912 | |||
BC075061.2 | - | 3 | 712 | 30915585 | ||
BC136424.1 | - | 3 | 909 | 9020413 |