Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other UBR4 SIRNA Assays ›
Gene Symbol:
UBR4
Gene Name:
ubiquitin protein ligase E3 component n-recognin 4
Gene Aliases:
RBAF600, ZUBR1, p600
Chromosome Location:
Chr.1: 19074506 - 19210334 on Build GRCh38
UniGene ID:
Hs.148078
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s23628
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: UBR4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_020765.2
NP_065816.2
46
6813
XM_011541108.2
XP_011539410.1
46
6913
XM_011541109.2
XP_011539411.1
46
6910
XM_011541110.2
XP_011539412.1
46
6913
XM_011541111.2
XP_011539413.1
46
6913
XM_011541112.2
XP_011539414.1
46
6898
XM_011541113.2
XP_011539415.1
46
6895
XM_011541114.2
XP_011539416.1
46
6913
XM_011541115.2
XP_011539417.1
46
6913
XM_011541116.2
XP_011539418.1
46
6913
XM_011541117.2
XP_011539419.1
46
6913
XM_011541118.2
XP_011539420.1
46
6913
XM_011541119.2
XP_011539421.1
46
6913
XM_011541120.2
XP_011539422.1
46
6913
XM_011541121.2
XP_011539423.1
46
6913
XM_017000822.1
XP_016856311.1
46
6913
XM_017000823.1
XP_016856312.1
46
6898
XM_017000824.1
XP_016856313.1
46
6913
XM_017000825.1
XP_016856314.1
46
6913
XM_017000826.1
XP_016856315.1
46
6895
XM_017000827.1
XP_016856316.1
46
6913
XM_017000828.1
XP_016856317.1
46
6895
XM_017000829.1
XP_016856318.1
46
6913
XM_017000830.1
XP_016856319.1
46
6895
GenBank mRNA
AB037728.1
-
29
4478
AF348492.1
-
46
6809
AK056218.1
-
6
950
BC015865.1
-
6
971
Back To Top
More Information
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Function(s)
Process(es)
viral process
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
ubiquitin-protein transferase activity
protein binding
calmodulin binding
zinc ion binding
ligase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline