Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RFWD2 SIRNA Assays ›
Gene Symbol:
RFWD2
Gene Name:
ring finger and WD repeat domain 2
Gene Aliases:
COP1, RNF200
Chromosome Location:
Chr.1: 175944826 - 176207279 on Build GRCh38
UniGene ID:
Hs.523744
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s34633
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RFWD2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001001740.3
NP_001001740.1
12
1646
NM_001286644.1
NP_001273573.1
11
1521
NM_022457.6
NP_071902.2
13
1718
XM_005245447.2
XP_005245504.1
13
1741
XM_005245448.2
XP_005245505.1
12
1693
XM_006711487.2
XP_006711550.1
12
1684
XM_011509881.1
XP_011508183.1
14
1807
XM_017002059.1
XP_016857548.1
14
1807
XM_017002060.1
XP_016857549.1
14
1795
XM_017002061.1
XP_016857550.1
13
1753
XM_017002062.1
XP_016857551.1
13
1747
XM_017002063.1
XP_016857552.1
13
1741
XM_017002064.1
XP_016857553.1
13
1738
XM_017002065.1
XP_016857554.1
12
1693
XM_017002066.1
XP_016857555.1
12
1684
XM_017002067.1
XP_016857556.1
12
1681
XM_017002068.1
XP_016857557.1
12
1678
XM_017002069.1
XP_016857558.1
8
1321
XM_017002070.1
XP_016857559.1
13
1513
XM_017002071.1
XP_016857560.1
11
902
XM_017002072.1
XP_016857561.1
11
900
XM_017002073.1
XP_016857562.1
10
842
XM_017002074.1
XP_016857563.1
10
840
XM_017002075.1
XP_016857564.1
10
922
XM_017002076.1
XP_016857565.1
11
905
XM_017002077.1
XP_016857566.1
9
697
XM_017002078.1
XP_016857567.1
11
970
XM_017002079.1
XP_016857568.1
11
1385
XM_017002080.1
XP_016857569.1
7
624
GenBank mRNA
AF508940.1
-
13
1452
AF527539.1
-
13
1715
AK001278.1
-
11
956
AK314750.1
-
13
1650
AY509921.1
-
12
1380
BC039723.1
-
11
968
BC094728.1
-
13
1706
5271629
KJ534928.1
-
13
1452
KJ534929.1
-
10
1257
KJ534930.1
-
12
1392
KJ534931.1
-
12
1383
KJ534932.1
-
13
1440
KJ534933.1
-
7
951
KJ535061.1
-
10
1157
KJ535076.1
-
12
1354
Back To Top
More Information
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Function(s)
Process(es)
response to ionizing radiation
protein ubiquitination
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
ubiquitin-protein transferase activity
protein binding
zinc ion binding
ligase activity
ubiquitin protein ligase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-nmj5c:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0