Search
Search

EFHC1| Gene Symbol: EFHC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172420.1 | NP_001165891.1 | 9 | 1538 | ||
| NM_018100.3 | NP_060570.2 | 8 | 1626 | |||
| NR_033327.1 | 7 | 2883 | ||||
| GenBank mRNA | AK001328.1 | - | 8 | 1512 | ||
| AK225543.1 | - | 7 | 2775 | |||
| AK295064.1 | - | 8 | 1369 | |||
| AK297632.1 | - | 9 | 1538 | |||
| AL122084.1 | - | 7 | 2769 | |||
| BC020210.1 | - | 8 | 1502 | 4586338 | ||
| EU520261.1 | - | 8 | 1511 | |||