Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FHL1 SIRNA Assays ›
Gene Symbol:
FHL1
Gene Name:
four and a half LIM domains 1
Gene Aliases:
FHL-1, FHL1A, FHL1B, FLH1A, KYOT, RBMX1A, RBMX1B, SLIM, SLIM-1, SLIM1, SLIMMER, XMPMA
Chromosome Location:
Chr.X: 136146702 - 136211359 on Build GRCh38
UniGene ID:
Hs.435369
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s5196
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FHL1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001159699.1
NP_001153171.1
4
643
NM_001159700.1
NP_001153172.1
5
638
NM_001159701.1
NP_001153173.1
4
514
NM_001159702.2
NP_001153174.1
5
707
NM_001159703.1
NP_001153175.1
5
707
NM_001159704.1
NP_001153176.1
4
1089
NM_001167819.1
NP_001161291.1
5
575
NM_001449.4
NP_001440.2
5
707
NR_027621.1
5
789
XM_006724743.2
XP_006724806.1
4
511
XM_006724744.2
XP_006724807.1
5
589
XM_006724745.3
XP_006724808.1
5
527
XM_006724746.2
XP_006724809.1
4
462
XM_006724747.2
XP_006724810.1
4
462
XM_011531316.1
XP_011529618.1
4
511
XM_017029357.1
XP_016884846.1
5
589
GenBank mRNA
AF063002.1
-
4
462
AF098518.1
-
4
431
AF220153.1
-
3
378
AK094024.1
-
3
223
AK122708.1
-
4
1090
AK223299.1
-
3
412
AK289411.1
-
5
789
AK297124.1
-
5
480
AK299379.1
-
4
369
AK299381.1
-
4
489
AK299450.1
-
5
423
AK300966.1
-
6
514
AK301642.1
-
4
511
AK304680.1
-
5
566
BC010998.2
-
5
586
4039973
BC088369.1
-
4
476
30529295
BE965488.2
-
5
515
CR456974.1
-
3
378
DA917361.1
-
3
273
DA982372.1
-
5
576
DC373304.1
-
4
515
U29538.1
-
3
399
U60115.1
-
4
461
U60118.1
-
2
64
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
regulation of membrane depolarization
muscle organ development
organ morphogenesis
negative regulation of G2/M transition of mitotic cell cycle
cell differentiation
negative regulation of cell growth
positive regulation of potassium ion transport
regulation of potassium ion transmembrane transporter activity
negative regulation of G1/S transition of mitotic cell cycle
molecular_function
protein binding
zinc ion binding
ion channel binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0