Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sntg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290390.1 | NP_001277319.1 | 19, 20 | 3261 | ||
NM_001290392.1 | NP_001277321.1 | 16, 17 | 2996 | |||
NM_001290393.1 | NP_001277322.1 | 18, 19 | 3210 | |||
NM_027671.5 | NP_081947.2 | 19, 20 | 3264 | |||
GenBank mRNA | AF367759.1 | - | 18, 19 | 1916 | ||
AK016927.1 | - | 5, 6 | 553 | |||
AK038698.1 | - | 16, 17 | 2997 | |||
AK080034.1 | - | 19, 20 | 2626 | |||
AK083241.1 | - | 19, 20 | 2249 |