Isolate and enrich for the transcriptome with Ribominus rRNA removal kits
Ribominus rRNA removal kits remove ribosomal RNA (rRNA) from your samples, and get them ready for next-generation sequencing or microarray. These rRNA depletion kits use biotin-labeled oligonucleotide probes to hybridize specifically to rRNA and streptavidin-coated magnetic beads to capture and remove the unwanted rRNA.
Order kits rRNA depletion overview
Features of RiboMinus rRNA depletion kits
  • Specific—species-specific probe design supports enrichment of coding and non-coding RNAs
  • Flexible—optimized kits available for both NGS and microarray-based analyses
  • Sensitive—provides increased representation of RNA transcript species relative to total RNA
  • Convenient—fits into your library prep workflow
Learn more about the Ribominus eukaryote rRNA depletion kit
RiboMinus rRNA depletion kit for bacteria
The Invitrogen RiboMinus Bacteria 2.0 Transcriptome Isolation Kit is designed to capture rRNA from a wide variety of prokaryotes
  • Accurate—optimized hybridization chemistry designed for whole transcriptome RNA-seq
  • Flexible—pan-prokaryote probes for bacterial rRNA depletion are compatible with dozens of species
  • High coverage—includes probes for 16S and 23S as well as 5S ribosomal RNA
  • Automation-compatible—can be used in manual or automated systems such as Thermo Scientific KingFisher Instruments
Design and performance of RiboMinus 2.0 Transcription kit
Learn more about the RiboMinus 2.0 Transcription kit works including list of targeted prokaryotes, rRNA depletion workflow. See the increase in bacterial transcriptome RNA-seq reads.
Download poster on bacterial rRNA depletion
View product details and FAQs
Ordering information for Ribominus rRNA depletion kits
Looking to do rRNA depletion on other species? Contact us
Contact us with your species and RNA sequence. Simply include your FASTA sequence in *.txt file format for the ribosomal sequences you would like to verify, with the subject line titled “RiboMinus for (your organism here),” and we'll work with you to find the right probe for your rRNA depletion work.
Stylesheet for Classic Wide Template adjustments
For Research Use Only. Not for use in diagnostic procedures.