Find valuable information.
Optimize your experiments to get the best results. We’ve compiled a detailed knowledge base of the top tips and tricks to meet your research needs.
View the relevant questions below:
Having problems with your experiment?  Visit our
Troubleshooting page
General
Salts, buffers, detergents, and other small molecules can interfere with liquid chromatography separation and mass spectrometer source ionization. 
For protein samples, Pierce Polyacrylamide Spin Desalting Columns (Cat. No. 89849 or 89862) can be used to remove salts and other small molecular weight contaminates. Acetone precipitation is recommended for low protein amounts provided you have at least 10 µg of sample. For peptide samples derived from protein digests, we recommend using Pierce Peptide Desalting Spin Columns, Pierce C18 Spin Tips or an in-line C18 trap column for sample clean-up.
No. Protein desalting columns (e.g., Zeba desalting columns) use size exclusion and have a molecular weight cut-off that is typically too high for peptide samples. Peptide desalting, on the other hand, uses reversed-phase chromatography resins (e.g., C18 resin) to bind peptides for salt removal during washing.
Most organic solvents or volatile buffers are compatible with LC-MS. Water and acetonitrile with 0.1% formic acid is the most common solvent system for separation of peptide samples for mass spectrometry analysis. Trifluoroacetic acid (TFA) is an alternative ion pairing agent for off-line peptide C18 clean-up or LC-UV analysis. We offer the following LC-MS grade reagents: 0.1% TFA, LC-MS water, acetonitrile, and TFA.
You can measure peptide concentration using our Pierce Quantitative Colorimetric Peptide Assay (Cat. No. 23275) or Pierce Quantitative Fluorometric Peptide Assay (Cat. No. 23290).
For Research Use Only. Not for use in diagnostic procedures.
CMD SchemaApp code