Search
We are dedicated to your success. Get back on track. View our expert recommendations for commonly encountered problem scenarios.
View the relevant questions below:
Beginning your experiment? Visit our
Follow the uninstall instructions for your version of GeneMapper Software. Before attempting to reinstall, verify that you are logged in locally as the Administrator, that your antivirus software is either off or that you have suspended activity during installation, and that your TCP/IP is installed.
Before uninstalling, make sure you have your GeneMapper CD/DVD and your registration code for reinstallation. Follow the uninstall instructions for your version of GeneMapper Software. Before attempting to reinstall, verify that you are logged in locally as the Administrator, that your antivirus Software is either off or that you have suspended activity during installation, and that your TCP/IP is installed.
If you received the error message about insufficient user privileges to start low level driver, you are most likely not logged in locally as the Administrator. Reboot the computer, change the domain to the local computer, and log in as the Administrator.
Right-click on Computer and select Properties. In the System section, look at the System Type and see if you have Windows 7 Professional, 32-bit or 64-bit edition. Write this down, as you will need this information during the install process.
To install the necessary driver:
Note: If your browser gives you the choice to Run or Save, save the file. For the Internet Explorer browser, you can choose the location to save the file. For Mozilla Firefox and Google Chrome browsers, the default location to save the file is the Downloads folder. 3. Run the installer. When the installation is complete, reboot the computer.
You should now be able to install the GeneMapper Software 5 Demo.
Right-click on Computer and select Properties. In the System section, look at the System Type and see if you have Windows 7 Professional, 32-bit or 64-bit edition. Write this down, as you will need this information during the install process.
To install the necessary driver:
Note: If your browser gives you the choice to Run or Save, save the file. For the Internet Explorer browser, you can choose the location to save the file. For Mozilla Firefox and Google Chrome browsers, the default location to save the file is the Downloads folder.
3. Run the installer. When the installation is complete, reboot the computer.
You should now be able to install the GeneMapper Software 5 Demo.
Please download this PDF (GeneMapper v.4.0 Uninstall– client install) and follow the steps listed to uninstall GeneMapper Software v.4.0 from your stand-alone computer.
Please download this PDF (GeneMapper v.4.0 uninstall) – full install and follow the steps listed to uninstall GeneMapper Software v.4.0.
Please download this PDF (GeneMapper v.4.1 Uninstall – client install) and follow the directions to uninstall GeneMapper Software v.4.1.
Please download this PDF (GeneMapper v.4.1 Uninstall Procedure – full install) and follow the directions to uninstall GeneMapper Software v.4.1.
Please download this PDF (GeneMapper 5 Uninstall – client install) and follow the directions to uninstall GeneMapper Software 5.
Please download this PDF (GeneMapper 5 Uninstall – full install) and follow the directions to uninstall GeneMapper Software 5.
Please download this PDF for directions on how to resolve this error.
Please download this PDF for directions on how to resolve this error.
Please download this PDF for directions on how to resolve this error.
It is useful to back up data in order to protect yourself from data loss, clearing out the Oracle database to keep it from filling up, and/or for sending data to Technical Support for troubleshooting.
In GeneMapper Software, there are 3 locations where you will need to back up data:
Note: Older versions of GeneMapper Software contained an Archive function that would back up the entire database at once. However, the archive could only be reimported into the same instance of GeneMapper Software. If you had a problem that required reinstallation of GeneMapper Software, it would be considered a new instance of the software and the Archive could not be imported into it.
Follow the steps outlined below to export Kits, Panels, Markers, and Bin Sets found in Panel Manager:
For backing up individual Panels or Bin Sets, click on the Kit (top-level folder) that contains the Panels and Markers you wish to back up. The File menu will now activate the options to Export Panels and Export Bin Sets.
To export project information, follow the steps outlined below:
When a GeneMapper Project is created, the sample files are not imported into GeneMapper Software, the location of the sample files is mapped within the project and edits are done to the project, leaving the raw data intact. This means that backing up a project will not also back up the files associated with the project. They will have to be backed up separately and reassociated to the project again if they are moved or if the software is reinstalled.
The location of the sample files will either be in the Runs folder (D:/AppliedBio/310/Runs) on the ABI PRISM 310 Genetic Analyzer system or it will be determined by the Results Group in the Data Collection Software on the multi-capillary platforms. If you do not know where that location is, you can go to the Data Collection Software for your instrument and go to the Results Group section (top left corner in Unified Data Collection; for 3500 Data Collection, go to Library and in the Manage section, select Results Group), open the Results Group, and check the Sample File Folder location.
Save the files to the storage media of your choice (CD, DVD, network drive, flash drive). If you are moving projects to a new system or reinstalling GeneMapper Software because of a hard drive failure, then you can move the sample files to the new location or new drive. When you open the project in GeneMapper Software, go to FileàAssociate Samples and browse to the folder containing the samples for the project.
See the associated GeneMapper Files, exported File Type, and data contained in each file below:
| File | Exported | Data Contained in File |
Projects | .ser | Sample file list, project settings, changes made to the project |
Analysis Methods | .xml | Analysis Settings and filters information |
Table Settings | .xml | What columns will be visible in the Samples and Genotypes table, font and size, and how it is sorted |
Plot Settings | .xml | Plot View Preferences. Appearance of tables in Sample plot and Genotypes plot, Sizing table, labels and Display Settings |
Cluster Plot Settings | .xml | Cluster Plot Preferences. SNP Table, Genotypes Cluster Plot table, and Display Settings for the Cluster Plot |
Matrices | .mtx | Matrix file for the ABI PRISM 310 Genetic Analyzer. Export to Matrix folder in the Data Collection Software |
Size Standards | .xml | Color of the standard and peak size (bp) information for peaks that will be called |
SNP Sets | .xml | SNPlex only. Information based on submitted SNP Pools |
Report Settings | .xml | Customize what information you want to appear on the analysis report |
Panels | .txt | Kit, Panel, and Marker information (color, range, name, etc.) |
Bin Sets | .txt | Bins associated with the Panels, specific Location of peaks within the marker range |
For Importing Data, you may want to start with the Panel Manager (Panels, Bin Sets) and then the GeneMapper Manager (Projects, Analysis Methods, etc.).
For Research Use Only. Not for use in diagnostic procedures.