19 Nov 2026
ASV_EDS
Collection of visualization modules for FIB-EDS data.
This module takes as input a stack of EM images and a set of stacks of EDS images and perform different image processing operations before displaying the data using different visualization techniques.
Known Limitations
- Fewer EDS slices can be acquired compared to SEM images.
- First and last slices must be acquired for each EDS channel.
- Distance between each EDS slice must be the same.
- Individual stack of EDS images will be resampled to match number of SEM slices and "missing" slices will be added using interpolation algorithm.
If you want to use this Xtra, please click on Request Information on this page. You will be driven to a contact form where you can inquire for a demo and/or commercial information and discussion.